Learn More
Eurofins Discoverx cAMP Hunter™ Human HTR1B (5-HT1B) Gi Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455836
Encompass
The cAMP Hunter™ Human HTR1B (5-HT1B) Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure HTR1B (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55836 Supplier Eurofins Discoverx Supplier No. 7950162C2
The cAMP Hunter™ Human HTR1B (5-HT1B) Gi Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure HTR1B (GPCR) activity via decreased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.