Learn More
Eurofins Discoverx cAMP Hunter™ Human HTR7 (5-HT7) Gs Cell-Based Assay Kit (DLD1)
Catalog No. 5000455982
Encompass
The cAMP Hunter™ Human HTR7 (5-HT7) Gs Stable Cell Line Assay (DLD1) contains a stable clonal cell line used to measure HTR7D (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55982 Supplier Eurofins Discoverx Supplier No. 7950163C13
The cAMP Hunter™ Human HTR7 (5-HT7) Gs Stable Cell Line Assay (DLD1) contains a stable clonal cell line used to measure HTR7D (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.