Learn More
Eurofins Discoverx cAMP Hunter™ Mouse GLP-1 (GLP-1 Receptor) Cell-Based Assay Kit (U2OS)
Catalog No. 5000456041
Encompass
The cAMP Hunter™ Mouse GLP-1 (GLP1R) Gs Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure mouse GLP1R (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56041 Supplier Eurofins Discoverx Supplier No. 7950179C3
The cAMP Hunter™ Mouse GLP-1 (GLP1R) Gs Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure mouse GLP1R (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.