Learn More
Eurofins Discoverx cAMP Hunter™ Human ADRB3 Gs Cell-Based Assay Kit (DLD1)
Catalog No. 5000455979
Encompass
The cAMP Hunter™ Human ADRB3 Gs Stable Cell Line Assay (DLD1) contains a stable clonal cell line used to measure ADRB3 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55979 Supplier Eurofins Discoverx Supplier No. 7950182C13
The cAMP Hunter™ Human ADRB3 Gs Stable Cell Line Assay (DLD1) contains a stable clonal cell line used to measure ADRB3 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.