Learn More
Eurofins Discoverx cAMP Hunter™ Mouse RXFP1 Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000456091
Encompass
The cAMP Hunter™ Mouse RXFP1 Gs Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure mouse RXFP1 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56091 Supplier Eurofins Discoverx Supplier No. 7950180C2
The cAMP Hunter™ Mouse RXFP1 Gs Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure mouse RXFP1 (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.