Learn More
Eurofins Discoverx FFAR1 (GPR40) Gq Calcium Signaling Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455848
Encompass
The FFAR1 (GPR40) Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure FFAR1 (GPR40) (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55848 Supplier Eurofins Discoverx Supplier No. 7951005C2
The FFAR1 (GPR40) Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure FFAR1 (GPR40) (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.