Learn More
Eurofins Discoverx F2RL1 Gq Calcium Signaling Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455849
Encompass
The F2RL1 Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure F2RL1 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55849 Supplier Eurofins Discoverx Supplier No. 7951006C2
The F2RL1 Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure F2RL1 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.