Learn More
Eurofins Discoverx ADORA2A (A2A) Gq Calcium Signaling Cell-Based Assay Kit (RBL-2H3)
Catalog No. 5000455506
Encompass
The ADORA2A (A2A) Gq Stable Cell Line Calcium Signaling Assay (RBL-2H3) contains a stable clonal cell line used to measure ADORA2A (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55506 Supplier Eurofins Discoverx Supplier No. 7951008C11
The ADORA2A (A2A) Gq Stable Cell Line Calcium Signaling Assay (RBL-2H3) contains a stable clonal cell line used to measure ADORA2A (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.