Learn More
Eurofins Discoverx S1P3 (EDG3) Gq Calcium Signaling Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455980
Encompass
The S1P3 (EDG3) Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure EDG3 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55980 Supplier Eurofins Discoverx Supplier No. 7951007C2
Add to Cart
Add to Cart
The S1P3 (EDG3) Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure EDG3 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.