Learn More
Eurofins Discoverx CYSLTR2 Gq Calcium Signaling Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000456052
Encompass
The CYSLTR2 Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure CYSLTR2 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56052 Supplier Eurofins Discoverx Supplier No. 7951013C2
The CYSLTR2 Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure CYSLTR2 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.