Learn More
Eurofins Discoverx GPR39 Gq Calcium Signaling Cell-Based Assay Kit (U2OS)
Catalog No. 5000456022
Encompass
The GPR39 Gq Stable Cell Line Calcium Signaling Assay (U2OS) contains a stable clonal cell line used to measure GPR39 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56022 Supplier Eurofins Discoverx Supplier No. 7951015C3
The GPR39 Gq Stable Cell Line Calcium Signaling Assay (U2OS) contains a stable clonal cell line used to measure GPR39 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.