Learn More
Eurofins Discoverx P2RY1 Gq Calcium Signaling Cell-Based Assay Kit (1321N1)
Catalog No. 5000455457
Encompass
The P2RY1 Gq Stable Cell Line Calcium Signaling Assay (1321N1) contains a stable clonal cell line used to measure P2RY1 (GPCR) activity via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55457 Supplier Eurofins Discoverx Supplier No. 7951018C6
The P2RY1 Gq Stable Cell Line Calcium Signaling Assay (1321N1) contains a stable clonal cell line used to measure P2RY1 (GPCR) activity via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.