Learn More
Eurofins Discoverx MRGPRX1 Gq Calcium Signaling Cell-Based Assay Kit (RBL-2H3)
Catalog No. 5000456106
Encompass
The MRGPRX1 Gq Stable Cell Line Calcium Signaling Assay (RBL-2H3) contains a stable clonal cell line used to measure MRGPRX1 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56106 Supplier Eurofins Discoverx Supplier No. 7951019C11
The MRGPRX1 Gq Stable Cell Line Calcium Signaling Assay (RBL-2H3) contains a stable clonal cell line used to measure MRGPRX1 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.