Learn More
Eurofins Discoverx GPR17 Gq Calcium Signaling Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455238
Encompass
The GPR17 Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure GPR17 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55238 Supplier Eurofins Discoverx Supplier No. 7951022C2
The GPR17 Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure GPR17 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.