Learn More
Eurofins Discoverx ADRA1A Gq Calcium Signaling Cell-Based Assay Kit (CHO-K1)
Supplier:  Eurofins Discoverx 7951023C2
Encompass
The ADRA1A Gq Stable Cell Line Calcium Signaling Assay (CHO-K1) contains a stable clonal cell line used to measure ADRA1A (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55315