Learn More
Eurofins Discoverx mGlu1 (GRM1) Gq Calcium Signaling Cell-Based Assay Kit (U2OS)
Catalog No. 5000455388
Encompass
The mGlu1 (GRM1) Gq Stable Cell Line Calcium Signaling Assay (U2OS) contains a stable clonal cell line used to measure GRM1 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55388 Supplier Eurofins Discoverx Supplier No. 7951021C3
The mGlu1 (GRM1) Gq Stable Cell Line Calcium Signaling Assay (U2OS) contains a stable clonal cell line used to measure GRM1 (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.