Learn More
Eurofins Discoverx HTR2B (5-HT2B) Gq Calcium Signaling Cell-Based Assay Kit (HEK 293)
Supplier:  Eurofins Discoverx 7951024C1
Encompass
The HTR2B (5-HT2B) Gq Stable Cell Line Calcium Signaling Assay (HEK 293) contains a stable clonal cell line used to measure HTR2B (GPCR) activation via an increase in intracellular calcium. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55405