Learn More
Eurofins Discoverx cAMP Hunter™ Human GLP-1 (GLP-1 Receptor) Gs Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455173
Encompass
The cAMP Hunter™ Human GLP-1 (GLP1R) Gs Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure GLP1R (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55173 Supplier Eurofins Discoverx Supplier No. 7951185C1
The cAMP Hunter™ Human GLP-1 (GLP1R) Gs Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure GLP1R (GPCR) activity via increased levels of intracellular cAMP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.