Learn More
Eurofins Discoverx InCELL Hunter™ Human G9a Methyltransferase Target Engagement Cell-Based Assay Kit (A549)
Catalog No. 5000455406
Encompass
The InCELL Hunter™ Human G9a Stable Cell Line Target Engagement Assay (A549) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of G9a target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55406 Supplier Eurofins Discoverx Supplier No. 7960003C15
The InCELL Hunter™ Human G9a Stable Cell Line Target Engagement Assay (A549) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of G9a target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.