Learn More
Eurofins Discoverx InCELL Hunter™ Human Brd2(1) Bromodomain Target Engagement Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455408
Encompass
The InCELL Hunter™ Human BRD2(1) Bromodomain Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of BRD2(1) target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55408 Supplier Eurofins Discoverx Supplier No. 7960004C1
The InCELL Hunter™ Human BRD2(1) Bromodomain Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of BRD2(1) target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.