Learn More
Eurofins Discoverx InCELL Hunter™ Human BRD3(1,2) Bromodomain Target Engagement Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455393
Encompass
The InCELL Hunter™ Human BRD3(1,2) Bromodomain Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of BRD3(1,2) target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55393 Supplier Eurofins Discoverx Supplier No. 7960038C1
The InCELL Hunter™ Human BRD3(1,2) Bromodomain Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of BRD3(1,2) target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.