Learn More
Eurofins Discoverx InCELL Hunter™ Human MEK1 Kinase Target Engagement Cell-Based Assay Kit (A549)
Catalog No. 5000455241
Encompass
The InCELL Hunter™ Human MEK1 Stable Cell Line Target Engagement Assay (A549) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of MEK1 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55241 Supplier Eurofins Discoverx Supplier No. 7960040C15
The InCELL Hunter™ Human MEK1 Stable Cell Line Target Engagement Assay (A549) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of MEK1 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.