Learn More
Eurofins Discoverx InCELL Hunter™ Human Blk Kinase Target Engagement Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455174
Encompass
The InCELL Hunter™ Human BLK Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of BLK target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55174 Supplier Eurofins Discoverx Supplier No. 7960044C1
The InCELL Hunter™ Human BLK Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of BLK target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.