Learn More
Eurofins Discoverx InCELL Hunter™ Human BTK Kinase Target Engagement Cell-Based Assay Kit (A549)
Catalog No. 5000455411
Encompass
The InCELL Hunter™ Human BTK Stable Cell Line Target Engagement Assay (A549) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of BTK target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55411 Supplier Eurofins Discoverx Supplier No. 7960056C15
The InCELL Hunter™ Human BTK Stable Cell Line Target Engagement Assay (A549) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of BTK target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.