Learn More
Eurofins Discoverx InCELL Hunter™ Human SMARCA4 Bromodomain Target Engagement Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455178
Encompass
The InCELL Hunter™ Human SMARCA4 Bromodomain Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of SMARCA4 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55178 Supplier Eurofins Discoverx Supplier No. 7960060C1
The InCELL Hunter™ Human SMARCA4 Bromodomain Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of SMARCA4 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.