Learn More
Eurofins Discoverx InCELL Hunter™ Human PIKFYVE Kinase Target Engagement Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455175
Encompass
The InCELL Hunter™ Human PIKFYVE Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of PIKFYVE target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55175 Supplier Eurofins Discoverx Supplier No. 7960078C1
The InCELL Hunter™ Human PIKFYVE Stable Cell Line Target Engagement Assay (HEK 293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of PIKFYVE target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.