Learn More
Eurofins Discoverx InCELL Hunter™ Human RIPK3 Kinase Target Engagement Cell-Based Assay Kit (Jurkat)
Catalog No. 5000455179
Encompass
The InCELL Hunter™ Human RIPK3 Stable Cell Line Target Engagement Assay (Jurkat) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of RIPK3 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55179 Supplier Eurofins Discoverx Supplier No. 7960077C19
Add to Cart
Add to Cart
The InCELL Hunter™ Human RIPK3 Stable Cell Line Target Engagement Assay (Jurkat) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of RIPK3 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.