Learn More
Eurofins Discoverx InCELL Hunter™ Human PKCθ Kinase Target Engagement Cell-Based Assay Kit (Jurkat)
Catalog No. 5000455394
Encompass
The InCELL Hunter™ Human PKC-theta Stable Cell Line Target Engagement Assay (Jurkat) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of PKC-theta target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55394 Supplier Eurofins Discoverx Supplier No. 7960082C19
The InCELL Hunter™ Human PKC-theta Stable Cell Line Target Engagement Assay (Jurkat) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of PKC-theta target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.