Learn More
Eurofins Discoverx InCELL Hunter™ Human ULK1 Kinase Target Engagement Cell-Based Assay Kit (HEK293)
Catalog No. 5000455410
Encompass
The InCELL Hunter™ Human ULK1 Stable Cell Line Target Engagement Assay (HEK293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of ULK1 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55410 Supplier Eurofins Discoverx Supplier No. 7960081C1
The InCELL Hunter™ Human ULK1 Stable Cell Line Target Engagement Assay (HEK293) contains a stable clonal cell line used to measure target engagement via compound-induced stabilization of ULK1 target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.