Learn More
Eurofins Discoverx KILR SKBR3 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455413
Encompass
The KILR SKBR3 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55413 Supplier Eurofins Discoverx Supplier No. 7971002P018
The KILR SKBR3 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.