Learn More
Eurofins Discoverx KILR ARH-77 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455416
Encompass
The KILR ARH-77 (Human) Stable Cell Line Cytotoxicity Assay utilizes the engineered stable target cell line that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55416 Supplier Eurofins Discoverx Supplier No. 7971001C017
The KILR ARH-77 (Human) Stable Cell Line Cytotoxicity Assay utilizes the engineered stable target cell line that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.