Learn More
Eurofins Discoverx KILR Ramos (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455248
Encompass
The KILR Ramos (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55248 Supplier Eurofins Discoverx Supplier No. 7971006P022
The KILR Ramos (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.