Learn More
Eurofins Discoverx KILR A549 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455415
Encompass
The KILR A549 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55415 Supplier Eurofins Discoverx Supplier No. 7971005P015
The KILR A549 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.