Learn More
Eurofins Discoverx KILR U2OS (Human) EGFR Cytotoxicity Assay Kit (Clone)
Catalog No. 5000455398
Encompass
The KILR Human EGFR Stable Cell Line Cytotoxicity Assay (U2OS) utilizes the stable, engineered U2OS-EGFR target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55398 Supplier Eurofins Discoverx Supplier No. 7971010C003
The KILR Human EGFR Stable Cell Line Cytotoxicity Assay (U2OS) utilizes the stable, engineered U2OS-EGFR target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.