Learn More
Eurofins Discoverx KILR MCF7 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455250
Encompass
The KILR MCF7 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55250 Supplier Eurofins Discoverx Supplier No. 7971014P028
The KILR MCF7 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.