Learn More
Eurofins Discoverx KILR U118-MG (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455185
Encompass
The KILR U118-MG (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55185 Supplier Eurofins Discoverx Supplier No. 7971018P032
The KILR U118-MG (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.