Learn More
Eurofins Discoverx KILRA498 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455252
Encompass
The KILRA498 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55252 Supplier Eurofins Discoverx Supplier No. 7971021P034
The KILRA498 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.