Learn More
Eurofins Discoverx KILR RPMI 8226 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455253
Encompass
The KILR RPMI 8226 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55253 Supplier Eurofins Discoverx Supplier No. 7971026P039
The KILR RPMI 8226 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.