Learn More
Eurofins Discoverx KILR U2OS (Human) PD-L1 Cytotoxicity Assay Kit (Clone)
Catalog No. 5000455401
Encompass
The KILR Human PD-L1 Stable Cell Line Cytotoxicity Assay (U2OS) utilizes the stable, engineered U2OS-PD-L1 target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55401 Supplier Eurofins Discoverx Supplier No. 7971029C003
The KILR Human PD-L1 Stable Cell Line Cytotoxicity Assay (U2OS) utilizes the stable, engineered U2OS-PD-L1 target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.