Learn More
Eurofins Discoverx KILR SK-MEL-5 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455323
Encompass
The KILR SK-MEL-5 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55323 Supplier Eurofins Discoverx Supplier No. 7971033P030
The KILR SK-MEL-5 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.