Learn More
Eurofins Discoverx KILR U2OS  (Human) PD-L2 Cytotoxicity Assay Kit (Clone)
Catalog No. 5000455255
Encompass
The KILR U2OS  Human PD-L2 Stable Cell Line Cytotoxicity Assay (U2OS) utilizes the stable, engineered U2OS-PD-L2 target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55255 Supplier Eurofins Discoverx Supplier No. 7971036C003
The KILR U2OS  Human PD-L2 Stable Cell Line Cytotoxicity Assay (U2OS) utilizes the stable, engineered U2OS-PD-L2 target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.