Learn More
Eurofins Discoverx KILR Jurkat (Human) LAG3 Cytotoxicity Assay Kit (Clone)
Catalog No. 5000455188
Encompass
The KILR Human LAG3 Stable Cell Line Cytotoxicity Assay (Jurkat) utilizes the stable, engineered Jurkat-LAG3 target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55188 Supplier Eurofins Discoverx Supplier No. 7971038C019
The KILR Human LAG3 Stable Cell Line Cytotoxicity Assay (Jurkat) utilizes the stable, engineered Jurkat-LAG3 target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.