Learn More
Eurofins Discoverx KILR HL-60 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455422
Encompass
The KILR HL-60 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55422 Supplier Eurofins Discoverx Supplier No. 7971039P046
The KILR HL-60 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct death mediated by immuno-oncology treatments via ADCC, CDC, and ADCP. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.