Learn More
Eurofins Discoverx KILR DU-145 (Human) Cytotoxicity Assay Kit (cell pool)
Catalog No. 5000455423
Encompass
The KILR DU-145 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct cell death via ADCC, CDC, and ADCP. This complete kit includes cell pool vials and all required reagents for cell culture, maintenance, and running the assay.
Catalog No. 50-004-55423 Supplier Eurofins Discoverx Supplier No. 7971043P050
The KILR DU-145 (Human) Stable Cell Pool Cytotoxicity Assay utilizes the engineered stable target cell pool that is used to measure direct cell death via ADCC, CDC, and ADCP. This complete kit includes cell pool vials and all required reagents for cell culture, maintenance, and running the assay.