Learn More
Eurofins Discoverx KILR Jurkat (Human) TIM3 Cytotoxicity Assay Kit (Clone)
Catalog No. 5000455404
Encompass
The KILR Human TIM3 Stable Cell Line Cytotoxicity Assay (Jurkat) utilizes the stable, engineered Jurkat-TIM3 target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55404 Supplier Eurofins Discoverx Supplier No. 7971046C019
The KILR Human TIM3 Stable Cell Line Cytotoxicity Assay (Jurkat) utilizes the stable, engineered Jurkat-TIM3 target cell line for quantifying effector-induced cell death. This assay enables measurement of immune cell-mediated killing through ADCC, ADCP and CDC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.