Learn More
Biochain Institute Inc ATP Assay Kit, 100; assays/PK
SDP
Catalog No. 50180507
Encompass_Preferred
BioChain's EnzyLightTM ATP Assay Kit provides a rapid method to measure intracellular ATP. The single working reagent lyses cells to release ATP, which, in the presence of luciferase, immediately reacts with the Substrate D-luciferin to produce light. The light intensity is a direct measure of intracellular ATPconcentration. This non-radioactive, homogeneous cell-based assay is performed in microplates. The reagent is compatible with all culture media and with all liquid handling systems for high throughput screning.
Catalog No. 50-180-507 Supplier Biochain Institute Inc Supplier No. Z5030041
BioChain's EnzyLightTM ATP Assay Kit provides a rapid method to measure intracellular ATP. The single working reagent lyses cells to release ATP, which, in the presence of luciferase, immediately reacts with the Substrate D-luciferin to produce light. The light intensity is a direct measure of intracellular ATPconcentration. This non-radioactive, homogeneous cell-based assay is performed in microplates. The reagent is compatible with all culture media and with all liquid handling systems for high throughput screning.