Learn More
ATCC Cell Basement Membrane
Catalog No. 502384011
Encompass
Cell Basement Membrane is purified from murine Engelbreth-Holm-Swarm (EHS) tumor. Cell Basement Membrane has applications in the three-dimensional culture of primary tissue-derived human and mouse organoids, a 3-D culture of iPSC-derived organoids, patient-derived organoids, a 3-D culture of tumorspheres from various cell lines, and a sandwich culture of human hepatocytes. Cell Basement Membrane is a feeder-free culture of human embryonic stem cells (hESC) and induced pluripotent stem cells (hiPSC), a culture of iPSCs-derived neural progenitor cells (NPCs) and neurons, and angiogenesis modeling via the tube formation assay.
Catalog No. 50-238-4011 Supplier ATCC Supplier No. ACS3035
Cell Basement Membrane is purified from murine Engelbreth-Holm-Swarm (EHS) tumor. Cell Basement Membrane has applications in the three-dimensional culture of primary tissue-derived human and mouse organoids, a 3-D culture of iPSC-derived organoids, patient-derived organoids, a 3-D culture of tumorspheres from various cell lines, and a sandwich culture of human hepatocytes. Cell Basement Membrane is a feeder-free culture of human embryonic stem cells (hESC) and induced pluripotent stem cells (hiPSC), a culture of iPSCs-derived neural progenitor cells (NPCs) and neurons, and angiogenesis modeling via the tube formation assay.