Learn More
Cell Biolabs Inc Urea Assay Kit, 200 assays
Catalog No. NC1785818
Encompass_Preferred
Cell Biolabs' Urea Assay Kit is a simple method for quantifying urea in a convenient 96-well microplate format. The kit detects urea levels as low as 0.78 mg/dL in plasma, serum, urine, cell lysates, or tissue homogenates. Each kit provides sufficient reagents for 200 assays including standards and unknown samples.
Catalog No. NC1785818 Supplier Cell Biolabs Inc Supplier No. STA382
Add to Cart
Add to Cart
Cell Biolabs' Urea Assay Kit is a simple method for quantifying urea in a convenient 96-well microplate format. The kit detects urea levels as low as 0.78 mg/dL in plasma, serum, urine, cell lysates, or tissue homogenates. Each kit provides sufficient reagents for 200 assays including standards and unknown samples.