Learn More
Apexbio Technology LLC cell-biology/cell-detection
SDP
Catalog No. 5000567620
Encompass_Preferred
Luminescent ATP Detection Assay Kit (Enhanced) is a key tool in biomedical research widely used for its ability to measure ATP levels with high sensitivity and precision Originating from advancements in bioluminescence technology this kit operates by targeting cellular ATP a critical energy molecule and quantifying its levels through a luminescent reaction The kit commonly demonstrates in vitro activity in the nanomolar to low micromolar range making it suitable for high-throughput screening It is typically applied in cell models drug screening and metabolic studies providing insights into cellular energy states and mitochondrial function Experimental concentrations typically depend on the specific research design allowing customization for varied scientific inquiries This assay offers researchers a reliable method to assess energy metabolism contributing to advances in understanding diseases and therapeutic development
Catalog No. 50-005-67620 Supplier Apexbio Technology LLC Supplier No. K2043200
Add to Cart
Add to Cart
Luminescent ATP Detection Assay Kit (Enhanced) is a key tool in biomedical research widely used for its ability to measure ATP levels with high sensitivity and precision Originating from advancements in bioluminescence technology this kit operates by targeting cellular ATP a critical energy molecule and quantifying its levels through a luminescent reaction The kit commonly demonstrates in vitro activity in the nanomolar to low micromolar range making it suitable for high-throughput screening It is typically applied in cell models drug screening and metabolic studies providing insights into cellular energy states and mitochondrial function Experimental concentrations typically depend on the specific research design allowing customization for varied scientific inquiries This assay offers researchers a reliable method to assess energy metabolism contributing to advances in understanding diseases and therapeutic development