Learn More
Alkali Scientific Alkali Scientific™ Cell Box Divider for 2" or 3" Cardboard Freezer Boxes, 196, 1/Pk
SDP
Catalog No. 503026402
Encompass_Preferred
Cardboard Material Perfect for Microtubes Additional box support and crush resistance Reusable and recyclable Makes packing and unpacking easy These economical cryo/freezer storage Cell Box Divider offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation. Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL Dimensions (L x W x H) 4 7/8 x 4 7/8 x 1 5/16″ (123.4 x 123.4 x 33mm)
Catalog No. 50-302-6402 Supplier Alkali Scientific Supplier No. CD196
Cardboard Material Perfect for Microtubes Additional box support and crush resistance Reusable and recyclable Makes packing and unpacking easy These economical cryo/freezer storage Cell Box Divider offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation. Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL Dimensions (L x W x H) 4 7/8 x 4 7/8 x 1 5/16″ (123.4 x 123.4 x 33mm)